Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pal Legionella pneu 50ug
SDP

Catalog No. 5000195989
Encompass_Preferred

Recombinant Pal protein from Legionella pneumophila, expressed in Escherichia coli with N-terminal SUMO and 6xHis tags. Supplied lyophilized from a PBS/trehalose solution to support storage stability, the protein is intended for biochemical and immunological studies of outer membrane integrity and Tol-Pal system function.

  • Recombinant protein expressed in Escherichia coli.
  • N-terminal SUMO and 6xHis tags for efficient purification.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight of ~36 kDa.
  • Lyophilized from PBS with 6% trehalose for improved stability.
  • Sequence provided (accession P26493, residues C22-R176) for sequence-specific applications.
  • Reconstitution guidance: do not reconstitute below 100 μg/mL in ddH2O.

Catalog No. 50-001-95989 Supplier Medchemexpress LLC Supplier No. HYP7146750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Pal protein from Legionella pneumophila, expressed in Escherichia coli with N-terminal SUMO and 6xHis tags. Supplied lyophilized from a PBS/trehalose solution to support storage stability, the protein is intended for biochemical and immunological studies of outer membrane integrity and Tol-Pal system function.

  • Recombinant protein expressed in Escherichia coli.
  • N-terminal SUMO and 6xHis tags for efficient purification.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight of ~36 kDa.
  • Lyophilized from PBS with 6% trehalose for improved stability.
  • Sequence provided (accession P26493, residues C22-R176) for sequence-specific applications.
  • Reconstitution guidance: do not reconstitute below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.