Learn More
Trajan Scientific Pal Microvial With Insert .3Ml
Catalog No. 501920895
Encompass
PAL System Micro-Vial 2CV 0.3mL Clear PP designed for the PAL Autosampler. 12x32mm with conical insert fits ND9 Screw Caps Pk of 100 Pcs
Catalog No. 501920895 Supplier Trajan Scientific Supplier No. VIAL0.3ND9PP100
PAL System Micro-Vial 2CV 0.3mL Clear PP designed for the PAL Autosampler. 12x32mm with conical insert fits ND9 Screw Caps Pk of 100 Pcs