Learn More
Malvern Panalytical PAL PELTIER STACK 6DW
Catalog No. NC3353835
Encompass
  • Maintains defined temperature conditions between 4 °C and 40 °C
  • Accommodates six standard microtiter plates, six deepwell plates, or six VT15, VT54, or VT70 racks
  • Simplifies setup with integrated power supply and cables
  • Supports automated laboratory workflows for sensitive samples
  • Designed for reproducibility and sample integrity
Catalog No. NC3353835 Supplier Malvern Panalytical Supplier No. GLT4006
Add to Cart
Add to Cart
Request a Quote
  • Maintains defined temperature conditions between 4 °C and 40 °C
  • Accommodates six standard microtiter plates, six deepwell plates, or six VT15, VT54, or VT70 racks
  • Simplifies setup with integrated power supply and cables
  • Supports automated laboratory workflows for sensitive samples
  • Designed for reproducibility and sample integrity