Learn More
Trajan Scientific Pal Screw Cap 18Mm Septa 1.5
Catalog No. 501920888
Encompass
PAL System Screw Cap 10CV and 20CV designed for the PAL Autosampler. ND18 Magnetic Silver Silicone white/PTFE blue Septa 1.5mm Pk of 100 pcs
Catalog No. 501920888 Supplier Trajan Scientific Supplier No. CAPND18STSP15100
PAL System Screw Cap 10CV and 20CV designed for the PAL Autosampler. ND18 Magnetic Silver Silicone white/PTFE blue Septa 1.5mm Pk of 100 pcs