Learn More
Trajan Scientific Pal Screw Cap 18Mm Septa 2.0
Catalog No. 501920889
Encompass
PAL System Screw Re-Cap 10CV and 20CV designed for the PAL Autosampler. ND18 Magnetic PTFE Metal Coating for optimized screw and sealing performance Silicone/PTFE Septa 2.0mm Pk of 100 pcs
Catalog No. 501920889 Supplier Trajan Scientific Supplier No. CAPND18STCSP20100
PAL System Screw Re-Cap 10CV and 20CV designed for the PAL Autosampler. ND18 Magnetic PTFE Metal Coating for optimized screw and sealing performance Silicone/PTFE Septa 2.0mm Pk of 100 pcs