Learn More
Trajan Scientific Pal Screw Cap 24Mm
Catalog No. 501920890
Encompass
PAL System Screw Cap 40CV designed for the PAL Autosampler. ND24 Silicone/PTFE Septa 3.2mm Pk of 100 pcs
Catalog No. 501920890 Supplier Trajan Scientific Supplier No. CAPND24PPSP32100
PAL System Screw Cap 40CV designed for the PAL Autosampler. ND24 Silicone/PTFE Septa 3.2mm Pk of 100 pcs