Learn More
Trajan Scientific Pal Screw Cap 9Mm Mag
Catalog No. 501920893
Encompass
PAL System Screw Cap 2CV designed for the PAL Autosampler. ND9 Magnetic Gold Silicone/PTFE Septa 1.0mm Pk of 100 pcs Similar to LAP.09151907
Catalog No. 501920893 Supplier Trajan Scientific Supplier No. CAPND9STSP10100
PAL System Screw Cap 2CV designed for the PAL Autosampler. ND9 Magnetic Gold Silicone/PTFE Septa 1.0mm Pk of 100 pcs Similar to LAP.09151907