Learn More
Trajan Scientific Pal Screw Cap 9Mm Pp Preslit
Catalog No. 501920892
Encompass
PAL System Screw Cap 2CV designed for the PAL Autosampler. ND9 Blue Silicone/PTFE/I-Slitted Septa 1.0mm Pk of 100 pcs
Catalog No. 501920892 Supplier Trajan Scientific Supplier No. CAPND9PPSP10S100
PAL System Screw Cap 2CV designed for the PAL Autosampler. ND9 Blue Silicone/PTFE/I-Slitted Septa 1.0mm Pk of 100 pcs