Learn More
Trajan Scientific Pal Snap Cap 20Mm
Catalog No. 501920887
Encompass
PAL System Clip Cap for PAL Wash Modules. 22mm PE with 8mm center hole Silicone/PTFE/Y-Slitted Septa 1.3mm Pk of 100 pcs
Catalog No. 501920887 Supplier Trajan Scientific Supplier No. CAPDC20PE8SP13Y100
PAL System Clip Cap for PAL Wash Modules. 22mm PE with 8mm center hole Silicone/PTFE/Y-Slitted Septa 1.3mm Pk of 100 pcs