Learn More
Cayman Chemical Pal-VGVAPG (acetate), Purity: ≥98%, 100 mg
SDP
Catalog No. 502911282
Encompass_Preferred
A lipidated hexapeptide composed of an elastin fragment conjugated to palmitic acid
Catalog No. 50-291-1282 Supplier Cayman Chemical Supplier No. 35185100 MG
A lipidated hexapeptide composed of an elastin fragment conjugated to palmitic acid