Learn More
Medchemexpress LLC Palacaparib | 2756333-39-6 | 98.3% | C21H22F2N6O2 | 10MG
SDP
Catalog No. 5000258189
Encompass_Preferred
Palacaparib is a CNS-penetrant, selective PARP1 inhibitor supplied for research use. It potently inhibits and traps PARP1 at sites of single-strand DNA breaks and is used in preclinical studies of homologous recombination-deficient cancers and other advanced solid malignancies.
  • CNS-penetrant PARP1 inhibitor.
  • High selectivity for PARP1 over other PARP family members.
  • Potent inhibition and trapping of PARP1 at single-strand breaks.
  • High purity (98.3%).
  • Supplied as a powder with defined cold-storage recommendations.
  • For research use only.
Catalog No. 50-002-58189 Supplier Medchemexpress LLC Supplier No. HY14580410MG
Palacaparib is a CNS-penetrant, selective PARP1 inhibitor supplied for research use. It potently inhibits and traps PARP1 at sites of single-strand DNA breaks and is used in preclinical studies of homologous recombination-deficient cancers and other advanced solid malignancies.
  • CNS-penetrant PARP1 inhibitor.
  • High selectivity for PARP1 over other PARP family members.
  • Potent inhibition and trapping of PARP1 at single-strand breaks.
  • High purity (98.3%).
  • Supplied as a powder with defined cold-storage recommendations.
  • For research use only.