Learn More
Aviva Systems Biology Corp PALB2 Antibody - N-terminal region (ARP63771_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63771_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PALB2. It was validated on Western Blot. . Alternate names: FANCN, PNCA3ApplicationWB. Species reactivity: Human
Catalog No. 50-288-078