Learn More
Medchemexpress LLC HY-50767 1g , Palbociclib PD 0332991 CAS:571190-30-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY507671G
Encompass_Preferred
Medchemexpress, HY-50767 1g Palbociclib PD 0332991 CAS:571190-30-2 Formula:C24H29N7O2 IC50: 11 nM (Cdk4), 16 nM (Cdk6) Purity:98% Palbociclib is a highly specific inhibitor of Cdk4 ( IC 50 =11 nM) and Cdk6 ( IC 50 =16 nM), having no activity against a panel of 36 additional protein kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2275