Learn More
Medchemexpress LLC HY-50767 500mg , Palbociclib CAS:571190-30-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50767500MG
Encompass_Preferred
HY-50767 500mg Palbociclib CAS: 571190-30-2 Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9207