Learn More
Selleck Chemical LLC Palbociclib S4482-25mg
SDP
Catalog No. 503108235
Encompass_Preferred
Palbociclib (PD 0332991) is a highly specific inhibitor of cyclin-dependent kinase 4 (Cdk4) and Cdk6 with IC50 of 11 nM and 16 nM respectively This product has poor solubility animal in vivo experiments are available cell experiments please choose carefully
Catalog No. 50-310-8235 Supplier Selleck Chemical LLC Supplier No. S448225MG
Palbociclib (PD 0332991) is a highly specific inhibitor of cyclin-dependent kinase 4 (Cdk4) and Cdk6 with IC50 of 11 nM and 16 nM respectively This product has poor solubility animal in vivo experiments are available cell experiments please choose carefully