Learn More
Medchemexpress LLC Palbociclib | 571190-30-2 | MFCD11840850 | 99.2% | 447.53 g/mol | C24H29N7O2 | 10 MG
SDP
Catalog No. 5000298620
Encompass_Preferred
Palbociclib analytical standard is the reference form of palbociclib (PD 0332991), a selective cyclin-dependent kinase 4 and 6 (CDK4/6) inhibitor. It is provided as a high-purity solid for research and analytical use, suitable for method development, qualitative and quantitative analysis by HPLC, GC, and MS.
  • High purity suitable for analytical and quantitative assays.
  • Intended for HPLC, GC, and MS method development and validation.
  • Solid form with light yellow to yellow appearance for easy handling.
  • Applicable to research on HR-positive, HER2-negative breast cancer models.
  • Provides a reliable reference for qualitative and quantitative measurements.
Catalog No. 50-002-98620 Supplier Medchemexpress LLC Supplier No. HY50767R10MG
Palbociclib analytical standard is the reference form of palbociclib (PD 0332991), a selective cyclin-dependent kinase 4 and 6 (CDK4/6) inhibitor. It is provided as a high-purity solid for research and analytical use, suitable for method development, qualitative and quantitative analysis by HPLC, GC, and MS.
  • High purity suitable for analytical and quantitative assays.
  • Intended for HPLC, GC, and MS method development and validation.
  • Solid form with light yellow to yellow appearance for easy handling.
  • Applicable to research on HR-positive, HER2-negative breast cancer models.
  • Provides a reliable reference for qualitative and quantitative measurements.