Learn More
Enzo Life Sciences Palbociclib (free base) (100mg). CAS: 571190-30-2
SDP
Supplier:  Enzo Life Sciences ENZCHM2320100
Encompass_Preferred
Palbociclib, also known as PD0332991, is an orally available pyridopyrimidine-derived cyclin-dependent kinase (CDK) inhibitor with potential antineoplastic activity. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3632