Learn More
Medchemexpress LLC Palbociclib hydrochloride | 571189-11-2 | 100.0% | C24H30ClN7O2 | 2 G
SDP
Catalog No. 5000359154
Encompass_Preferred
Palbociclib hydrochloride is an orally active, selective inhibitor of CDK4 and CDK6, with IC50 values of 11 nM and 16 nM respectively. It demonstrates potent anti-proliferative activity and is known to induce cell cycle arrest in various cancer cells. This compound is suitable for research in HR-positive and HER2-negative breast cancer, as well as hepatocellular carcinoma.
  • Orally active selective CDK4 and CDK6 inhibitor
  • Potent anti-proliferative activity
  • Induces cell cycle arrest in cancer cells
  • Used in research for HR-positive and HER2-negative breast cancer and hepatocellular carcinoma
Catalog No. 50-003-59154 Supplier Medchemexpress LLC Supplier No. HY50767C2G
Palbociclib hydrochloride is an orally active, selective inhibitor of CDK4 and CDK6, with IC50 values of 11 nM and 16 nM respectively. It demonstrates potent anti-proliferative activity and is known to induce cell cycle arrest in various cancer cells. This compound is suitable for research in HR-positive and HER2-negative breast cancer, as well as hepatocellular carcinoma.
  • Orally active selective CDK4 and CDK6 inhibitor
  • Potent anti-proliferative activity
  • Induces cell cycle arrest in cancer cells
  • Used in research for HR-positive and HER2-negative breast cancer and hepatocellular carcinoma