Learn More
Medchemexpress LLC Palbociclib hydrochloride | 571189-11-2 | 99.96% | C24H30ClN7O2 | 50 MG
SDP
Catalog No. 5000357905
Encompass_Preferred
Palbociclib hydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. It exhibits potent anti-proliferative activity and induces cell cycle arrest in cancer cells, making it suitable for research in HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
  • Orally active selective CDK4 and CDK6 inhibitor
  • Exhibits potent anti-proliferative activity
  • Induces cell cycle arrest in cancer cells
  • Suitable for research in HR-positive and HER2-negative breast cancer
  • Suitable for research in hepatocellular carcinoma
Catalog No. 50-003-57905 Supplier Medchemexpress LLC Supplier No. HY50767C50MG
Palbociclib hydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. It exhibits potent anti-proliferative activity and induces cell cycle arrest in cancer cells, making it suitable for research in HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
  • Orally active selective CDK4 and CDK6 inhibitor
  • Exhibits potent anti-proliferative activity
  • Induces cell cycle arrest in cancer cells
  • Suitable for research in HR-positive and HER2-negative breast cancer
  • Suitable for research in hepatocellular carcinoma