Learn More
Medchemexpress LLC Palbociclib hydrochloride (PD-0332991 hydrochloride) | 571189-11-2 | 100.0% | 500 MG
SDP
Catalog No. 5000358337
Encompass_Preferred
Palbociclib (PD 0332991) hydrochloride is an orally active, selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. It exhibits potent anti-proliferative activity and is known to induce cell cycle arrest in cancer cells. This compound can be utilized in research related to HR-positive and HER2-negative breast cancer, as well as hepatocellular carcinoma.
  • Orally active selective CDK4 and CDK6 inhibitor.
  • Exhibits potent anti-proliferative activity.
  • Induces cell cycle arrest in cancer cells.
  • Can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
  • Inhibits retinoblastoma phosphorylation at Ser795.
  • Arrests MDA-MB-453 cells exclusively in G1 phase.
Catalog No. 50-003-58337 Supplier Medchemexpress LLC Supplier No. HY50767C500MG
Palbociclib (PD 0332991) hydrochloride is an orally active, selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. It exhibits potent anti-proliferative activity and is known to induce cell cycle arrest in cancer cells. This compound can be utilized in research related to HR-positive and HER2-negative breast cancer, as well as hepatocellular carcinoma.
  • Orally active selective CDK4 and CDK6 inhibitor.
  • Exhibits potent anti-proliferative activity.
  • Induces cell cycle arrest in cancer cells.
  • Can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
  • Inhibits retinoblastoma phosphorylation at Ser795.
  • Arrests MDA-MB-453 cells exclusively in G1 phase.