Learn More
Medchemexpress LLC Palbociclib impurity 20 | 1774898-55-3 | 99.9% | 308.34 | 5 MG
SDP
Catalog No. 5000404014
Encompass_Preferred
Palbociclib impurity 20 is an impurity of Palbociclib (HY-50767). It has a CAS No. of 1774898-55-3 and a Molecular Weight of 308.34. The chemical formula is C14H20N4O4 and it appears as a solid.
  • High purity for reliable research
  • Shipped at room temperature in continental US
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-04014 Supplier Medchemexpress LLC Supplier No. HY429255MG
Palbociclib impurity 20 is an impurity of Palbociclib (HY-50767). It has a CAS No. of 1774898-55-3 and a Molecular Weight of 308.34. The chemical formula is C14H20N4O4 and it appears as a solid.
  • High purity for reliable research
  • Shipped at room temperature in continental US
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month