Learn More
Medchemexpress LLC Palbociclib monohydrochloride | 827022-32-2 | 99.98% | C24H30ClN7O2 | 2 G
SDP
Supplier:  Medchemexpress LLC HY50767A2G
Encompass_Preferred
Palbociclib monohydrochloride (PD 0332991) is an orally active, selective inhibitor of CDK4 and CDK6. It demonstrates potent anti-proliferative activity and induces cell cycle arrest in cancer cells. This compound is utilized in research for HR-positive and HER2-negative breast cancer, as well as hepatocellular carcinoma.
  • Orally active, selective inhibitor of CDK4 and CDK6.
  • Demonstrates potent anti-proliferative activity.
  • Induces cell cycle arrest in cancer cells.
  • Utilized in research for HR-positive and HER2-negative breast cancer.
  • Used in research for hepatocellular carcinoma.
Catalog No. 50-003-59305