Learn More
Aldevron pALD-AAV2 10mg
Catalog No. 503073864
Encompass_Preferred
10mg dispense of AAV2 RepCap plasmid for use in an AAV triple transfection.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3864 Supplier Aldevron Supplier No. 505710
10mg dispense of AAV2 RepCap plasmid for use in an AAV triple transfection.
This product is subject to the following: Limited Use License