Learn More
Aldevron pALD-AAV6 10mg
Catalog No. 503073890
Encompass_Preferred
10mg dispense of AAV6 RepCap plasmid for use in an AAV triple transfection.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3890 Supplier Aldevron Supplier No. 505910
10mg dispense of AAV6 RepCap plasmid for use in an AAV triple transfection.
This product is subject to the following: Limited Use License