Learn More
Aldevron pALD-CV77-Luciferase 10mg
Catalog No. 503073874
Encompass_Preferred
10mg dispense of the pALD-CV77 Luciferase reporter plasmid. Consistent with the pALD-CV77 cloning backbone, Aldevron offers a pre-made reporter construct, pALD-CV77-Luciferase, which has several advantages:
Commercially available sequence for research through clinical use
Kanamycin resistance
Consistent, reliable supply
Immediately available at research grade
This product is subject to the following: Limited Use License
Catalog No. 50-307-3874 Supplier Aldevron Supplier No. 507810
10mg dispense of the pALD-CV77 Luciferase reporter plasmid. Consistent with the pALD-CV77 cloning backbone, Aldevron offers a pre-made reporter construct, pALD-CV77-Luciferase, which has several advantages:
Commercially available sequence for research through clinical use
Kanamycin resistance
Consistent, reliable supply
Immediately available at research grade
This product is subject to the following: Limited Use License