Learn More
Aldevron pALD-ITR-GFP 10mg
Catalog No. 503073891
Encompass_Preferred
10mg dispense of GFP reporter version of the pALD-AAV system suitable for testing and optimization.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3891 Supplier Aldevron Supplier No. 506210
10mg dispense of GFP reporter version of the pALD-AAV system suitable for testing and optimization.
This product is subject to the following: Limited Use License