Learn More
Aldevron pALD-ITR-WPRE-GFP 10mg
Catalog No. 503073865
Encompass_Preferred
GFP reporter version of the pALD-AAV system suitable for testing and optimization. Contains WPRE sequence, 10mg dispense.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3865 Supplier Aldevron Supplier No. 506910
GFP reporter version of the pALD-AAV system suitable for testing and optimization. Contains WPRE sequence, 10mg dispense.
This product is subject to the following: Limited Use License