Learn More
Aldevron pALD-X80 10mg
Catalog No. 503073884
Encompass_Preferred
10mg dispense of pALD-X80 helper plasmid. 18.9kb, Kanamycin resistant, license and royalty free from research through commercial applications. pALD-X80 has a history of use in clinical trials, and has a Drug Master File on file with the US FDA.
This product is subject to the following: Limited Use License
Catalog No. 50-307-3884 Supplier Aldevron Supplier No. 501710
10mg dispense of pALD-X80 helper plasmid. 18.9kb, Kanamycin resistant, license and royalty free from research through commercial applications. pALD-X80 has a history of use in clinical trials, and has a Drug Master File on file with the US FDA.
This product is subject to the following: Limited Use License