Learn More
Cayman Chemical Palifosfamide, Purity: ≥95%, CAS: 31645-39-3, 100 mg
SDP
Catalog No. 502990616
Encompass_Preferred
An active metabolite of ifosfamide; formed through hydroxylation of ifosfamide by various hepatic CYP isoforms; cytotoxic to L1210 and CCRF-CEM cancer cells at 100 µM; reduces tumor growth in Lewis lung carcinoma and L1210 leukemia mouse models, as well as a rat Yoshida ascitic sarcoma model at 100 mg/kg per da
Catalog No. 50-299-0616 Supplier Cayman Chemical Supplier No. 29887100 MG
An active metabolite of ifosfamide; formed through hydroxylation of ifosfamide by various hepatic CYP isoforms; cytotoxic to L1210 and CCRF-CEM cancer cells at 100 µM; reduces tumor growth in Lewis lung carcinoma and L1210 leukemia mouse models, as well as a rat Yoshida ascitic sarcoma model at 100 mg/kg per da