Learn More
Apexbio Technology LLC Palifosfamide 31645-39-3 200mg
SDP
Catalog No. 5000062859
Encompass_Preferred
Palifosfamide (CAS 31645-39-3) is the active metabolite derivative of ifosfamide (IFA) an alkylating agent exhibiting cytotoxic effects through cross-linking DNA strands thereby interfering with replication and transcription Palifosfamide lysine demonstrates anticancer activity against pediatric sarcoma cell lines including osteosarcoma models such as SAOS-2 (IC50 2 25 6 75 M) and decreased sensitivity in OS222 (IC50 31 5 M) In murine xenograft experiments (MX-1 tumors) palifosfamide treatment at 100 mg/kg showed substantial tumor growth inhibition ( 80%) and partial complete response rates ( 17%) highlighting its applicability in preclinical oncology research
Catalog No. 50-000-62859 Supplier Apexbio Technology LLC Supplier No. A3696200
Palifosfamide (CAS 31645-39-3) is the active metabolite derivative of ifosfamide (IFA) an alkylating agent exhibiting cytotoxic effects through cross-linking DNA strands thereby interfering with replication and transcription Palifosfamide lysine demonstrates anticancer activity against pediatric sarcoma cell lines including osteosarcoma models such as SAOS-2 (IC50 2 25 6 75 M) and decreased sensitivity in OS222 (IC50 31 5 M) In murine xenograft experiments (MX-1 tumors) palifosfamide treatment at 100 mg/kg showed substantial tumor growth inhibition ( 80%) and partial complete response rates ( 17%) highlighting its applicability in preclinical oncology research