Learn More
Cayman Chemical PalIperIdon 50mg
SDP
Catalog No. 502973026
Encompass_Preferred
An atypical antipsychotic; an inverse agonist of dopamine D2 and D3 receptors (IC50s = 1 and 0.9 nM, respectively) as well as an antagonist at these receptors (Kis = 0.4 and 0.1 nM, respectively); binds to α1- and α2-adrenergic receptors, histamine H1 receptors, and the 5-HT receptor subtypes 5-HT1A , 5-HT1D, 5-HT2A, and 5-HT2C (Kds = 10.1, 80, 3.4, 480, 19, 1.21, and 48 nM, respectively); improves spatial working memory in a T-maze alternation task in a maternal immune activation mouse model of schizophrenia at 0.05 mg/kg per day; an active metabolite of risperidone
Catalog No. 50-297-3026 Supplier Cayman Chemical Supplier No. 1555650 MG
An atypical antipsychotic; an inverse agonist of dopamine D2 and D3 receptors (IC50s = 1 and 0.9 nM, respectively) as well as an antagonist at these receptors (Kis = 0.4 and 0.1 nM, respectively); binds to α1- and α2-adrenergic receptors, histamine H1 receptors, and the 5-HT receptor subtypes 5-HT1A , 5-HT1D, 5-HT2A, and 5-HT2C (Kds = 10.1, 80, 3.4, 480, 19, 1.21, and 48 nM, respectively); improves spatial working memory in a T-maze alternation task in a maternal immune activation mouse model of schizophrenia at 0.05 mg/kg per day; an active metabolite of risperidone