Learn More
Cayman Chemical Paliperidone-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1020719-55-4, 1 mg
SDP
Catalog No. 502938335
Encompass_Preferred
An internal standard for the quantification of paliperidone by GC- or LC-MS
Catalog No. 50-293-8335 Supplier Cayman Chemical Supplier No. 284841 MG
An internal standard for the quantification of paliperidone by GC- or LC-MS