Learn More
Cayman Chemical PalIperIdon-d4 500ug
SDP
Supplier:  Cayman Chemical 28484500 UG
Encompass_Preferred
An internal standard for the quantification of paliperidone by GC- or LC-MS
Catalog No. 50-293-9079