Learn More
Medchemexpress LLC Paliperidone (9-Hydroxyrisperidone) | 144598-75-4 | 99.4% | C23H27FN4O3 | 200 MG
SDP
Catalog No. 5000433074
Encompass_Preferred
Paliperidone (9-Hydroxyrisperidone) is the major active metabolite of Risperidone, functioning as a dopamine D2 and 5-HT2A antagonist. It also exhibits antagonist activity at α1 and α2 adrenergic receptors and H1-histaminergic receptors, demonstrating efficacy as an antipsychotic agent against schizophrenia.
  • Major active metabolite of risperidone
  • Dopamine D2 antagonist
  • 5-HT2A antagonist
  • Antagonist at α1 and α2 adrenergic receptors
  • Antagonist at H1-histaminergic receptors
  • Effective antipsychotic agent against schizophrenia
  • For research use only
  • Increases caspase-3 activity in neuroblastoma cells
  • May modulate Akt1/GSK3β pathway to protect SK-N-SH cells from glutamate damage
  • Can protect SK-N-SH cells from apoptosis induced by glutamate
Catalog No. 50-004-33074 Supplier Medchemexpress LLC Supplier No. HYA0019200MG
Paliperidone (9-Hydroxyrisperidone) is the major active metabolite of Risperidone, functioning as a dopamine D2 and 5-HT2A antagonist. It also exhibits antagonist activity at α1 and α2 adrenergic receptors and H1-histaminergic receptors, demonstrating efficacy as an antipsychotic agent against schizophrenia.
  • Major active metabolite of risperidone
  • Dopamine D2 antagonist
  • 5-HT2A antagonist
  • Antagonist at α1 and α2 adrenergic receptors
  • Antagonist at H1-histaminergic receptors
  • Effective antipsychotic agent against schizophrenia
  • For research use only
  • Increases caspase-3 activity in neuroblastoma cells
  • May modulate Akt1/GSK3β pathway to protect SK-N-SH cells from glutamate damage
  • Can protect SK-N-SH cells from apoptosis induced by glutamate