Learn More
Selleck Chemical LLC Paliperidone 25mg 144598-75-4
SDP
Catalog No. 506119
Encompass_Preferred
Paliperidone, the main active metabolite of Risperidone, is a potent serotonin-2A and dopamine-2 receptor antagonist, used in the treatment of schizophrenia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-611-9 Supplier Selleck Chemical LLC Supplier No. S172425MG
Paliperidone, the main active metabolite of Risperidone, is a potent serotonin-2A and dopamine-2 receptor antagonist, used in the treatment of schizophrenia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.