Learn More
Medchemexpress LLC Paliperidone palmitate | 199739-10-1 | 99.9% | C39H57FN4O4 | 100 MG
SDP
Catalog No. 5000433224
Encompass_Preferred
Paliperidone palmitate is an orally effective competitive antagonist of dopamine D2 receptors and 5-hydroxytryptamine 2A (5-HT2A) receptors that can cross the blood-brain barrier. It competitively inhibits the effects of dopamine and 5-hydroxytryptamine by binding to dopamine D2 receptors and 5-HT2A receptors, regulating the balance of the neurotransmitter system and thus exerting antipsychotic activity. It is mainly used in the research field of schizophrenia.
  • White to off-white solid appearance
  • Molecular weight: 664.89
  • Competitively inhibits dopamine D2 and 5-HT2A receptors
  • Regulates neurotransmitter system balance
  • Exerts antipsychotic activity
  • Used in schizophrenia research
Catalog No. 50-004-33224 Supplier Medchemexpress LLC Supplier No. HYA0019A100MG
Paliperidone palmitate is an orally effective competitive antagonist of dopamine D2 receptors and 5-hydroxytryptamine 2A (5-HT2A) receptors that can cross the blood-brain barrier. It competitively inhibits the effects of dopamine and 5-hydroxytryptamine by binding to dopamine D2 receptors and 5-HT2A receptors, regulating the balance of the neurotransmitter system and thus exerting antipsychotic activity. It is mainly used in the research field of schizophrenia.
  • White to off-white solid appearance
  • Molecular weight: 664.89
  • Competitively inhibits dopamine D2 and 5-HT2A receptors
  • Regulates neurotransmitter system balance
  • Exerts antipsychotic activity
  • Used in schizophrenia research