Learn More
Medchemexpress LLC Paliperidone palmitate | 199739-10-1 | 99.9% | C39H57FN4O4 | 25 MG
SDP
Catalog No. 5000358828
Encompass_Preferred
Paliperidone palmitate is an orally effective competitive antagonist of dopamine D2 receptors and 5-hydroxytryptamine 2A (5-HT2A) receptors that can cross the blood-brain barrier. It competitively inhibits the effects of dopamine and 5-hydroxytryptamine by binding to these receptors, regulating the balance of the neurotransmitter system and thus exerting antipsychotic activity. It is primarily utilized in the research of schizophrenia.
  • Competitive antagonist for dopamine D2 and 5-HT2A receptors.
  • Crosses the blood-brain barrier.
  • Regulates neurotransmitter system balance.
  • Exerts antipsychotic activity.
  • Targets dopamine and 5-HT receptors.
  • Involved in GPCR/G Protein and neuronal signaling pathways.
  • Used in schizophrenia research.
Catalog No. 50-003-58828 Supplier Medchemexpress LLC Supplier No. 25MGHYA0019A
Paliperidone palmitate is an orally effective competitive antagonist of dopamine D2 receptors and 5-hydroxytryptamine 2A (5-HT2A) receptors that can cross the blood-brain barrier. It competitively inhibits the effects of dopamine and 5-hydroxytryptamine by binding to these receptors, regulating the balance of the neurotransmitter system and thus exerting antipsychotic activity. It is primarily utilized in the research of schizophrenia.
  • Competitive antagonist for dopamine D2 and 5-HT2A receptors.
  • Crosses the blood-brain barrier.
  • Regulates neurotransmitter system balance.
  • Exerts antipsychotic activity.
  • Targets dopamine and 5-HT receptors.
  • Involved in GPCR/G Protein and neuronal signaling pathways.
  • Used in schizophrenia research.