Learn More
Medchemexpress LLC Paliperidone palmitate | 199739-10-1 | 99.9% | C39H57FN4O4 | 50 MG
SDP
Catalog No. 5000432900
Encompass_Preferred
Paliperidone palmitate is an orally effective competitive antagonist of dopamine D2 and 5-hydroxytryptamine 2A (5-HT2A) receptors, capable of crossing the blood-brain barrier. It regulates neurotransmitter system balance by inhibiting dopamine and 5-hydroxytryptamine effects, thereby exerting antipsychotic activity. This compound is primarily utilized in schizophrenia research.
  • Orally effective competitive antagonist of dopamine D2 receptors
  • Antagonist of 5-hydroxytryptamine 2A (5-HT2A) receptors
  • Crosses the blood-brain barrier
  • Inhibits dopamine and 5-hydroxytryptamine effects
  • Regulates neurotransmitter system balance
  • Exerts antipsychotic activity
  • Used in schizophrenia research
Catalog No. 50-004-32900 Supplier Medchemexpress LLC Supplier No. HYA0019A50MG
Paliperidone palmitate is an orally effective competitive antagonist of dopamine D2 and 5-hydroxytryptamine 2A (5-HT2A) receptors, capable of crossing the blood-brain barrier. It regulates neurotransmitter system balance by inhibiting dopamine and 5-hydroxytryptamine effects, thereby exerting antipsychotic activity. This compound is primarily utilized in schizophrenia research.
  • Orally effective competitive antagonist of dopamine D2 receptors
  • Antagonist of 5-hydroxytryptamine 2A (5-HT2A) receptors
  • Crosses the blood-brain barrier
  • Inhibits dopamine and 5-hydroxytryptamine effects
  • Regulates neurotransmitter system balance
  • Exerts antipsychotic activity
  • Used in schizophrenia research