Learn More
AdipoGen Palitantin (1 mg)
SDP
Catalog No. 502053517
Encompass_Preferred
Chemical. CAS: 15265-28-8. Formula: C14H22O4. MW: 254.3. Isolated from Penicillium sp. Antiprotozoal (Leishmania). Antifungal.
Catalog No. 50-205-3517 Supplier AdipoGen Supplier No. AGCN20140M001
Chemical. CAS: 15265-28-8. Formula: C14H22O4. MW: 254.3. Isolated from Penicillium sp. Antiprotozoal (Leishmania). Antifungal.