Learn More
Andwin Scientific PALL SOLVAC FILTER HOLDER 1/EA
SDP
Supplier:  Andwin Scientific 4020
Encompass_Preferred
The Solvac filter holders remove contaminating particulates and degas mobile phase solvents up to 4L. It features polypropylene construction for ideal chemical compatibility and magnetic seal.
Catalog No. NC0661821