Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 10g. Palladium, 10% on activated carbon, reduced, dry powder. MFCD03457879
SDP
Catalog No. 5090112399
Encompass
CAS# 7440-05-3. 10g. Palladium, 10% on activated carbon, reduced, dry powder. MFCD03457879. Molecular Weight: . Molecular Formula: Pd on carbon. Color/form: pwdr. Strem# 46-1900. http://www.strem.com/catalog/v/46-1900/
Catalog No. 50-901-12399 Supplier Strem, An Ascensus Company Supplier No. 46190010G
CAS# 7440-05-3. 10g. Palladium, 10% on activated carbon, reduced, dry powder. MFCD03457879. Molecular Weight: . Molecular Formula: Pd on carbon. Color/form: pwdr. Strem# 46-1900. http://www.strem.com/catalog/v/46-1900/