Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 10g. Palladium, 10% on activated wood carbon, reduced, 50% water wet (Escat™ 1931). MFCD03457879
SDP
Catalog No. 5090112409
Encompass
CAS# 7440-05-3. 10g. Palladium, 10% on activated wood carbon, reduced, 50% water wet (Escat™ 1931). MFCD03457879. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=38 mum) Strem# 46-1905. http://www.strem.com/catalog/v/46-1905/
Catalog No. 50-901-12409 Supplier Strem, An Ascensus Company Supplier No. 46190510G
CAS# 7440-05-3. 10g. Palladium, 10% on activated wood carbon, reduced, 50% water wet (Escat™ 1931). MFCD03457879. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=38 mum) Strem# 46-1905. http://www.strem.com/catalog/v/46-1905/