Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 10g. Palladium, 10% on activated wood carbon, unreduced, 50% water wet (Escat™ 1921). MFCD03457879
SDP
Catalog No. 5090112411
Encompass
CAS# 7440-05-3. 10g. Palladium, 10% on activated wood carbon, unreduced, 50% water wet (Escat™ 1921). MFCD03457879. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=38 mum) Strem# 46-1906. http://www.strem.com/catalog/v/46-1906/
Catalog No. 50-901-12411 Supplier Strem, An Ascensus Company Supplier No. 46190610G
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CAS# 7440-05-3. 10g. Palladium, 10% on activated wood carbon, unreduced, 50% water wet (Escat™ 1921). MFCD03457879. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=38 mum) Strem# 46-1906. http://www.strem.com/catalog/v/46-1906/