Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium, 5% on barium sulfate powder, reduced

Catalog No. AA1172214
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1172214 25 g
AA1172288 2.5 g
AA1172222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1172214 Supplier Thermo Scientific Chemicals Supplier No. 01172214
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-05-3 | Pd | 106.42 g/mol

Palladium, 5% on barium sulfate powder reduced usually supported on Barium sulfate, or an appropriate form of carbon, and useful catalyst for hydrogenations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Gray to Black
Appearance grayish black powder
Quantity 25 g
Odor Odorless
Form Powder
Solubility Information Insoluble in water.
Chemical Name or Material Palladium, 5% on barium sulfate powder
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.