Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 10g. Palladium, 5% on activated wood carbon, reduced, 50% water wet paste (Escat™ 1421). MFCD03457879
SDP
Catalog No. 5090112405
Encompass
CAS# 7440-05-3. 10g. Palladium, 5% on activated wood carbon, reduced, 50% water wet paste (Escat™ 1421). MFCD03457879. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=18 mum) Strem# 46-1903. http://www.strem.com/catalog/v/46-1903/
Catalog No. 50-901-12405 Supplier Strem, An Ascensus Company Supplier No. 46190310G
CAS# 7440-05-3. 10g. Palladium, 5% on activated wood carbon, reduced, 50% water wet paste (Escat™ 1421). MFCD03457879. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=18 mum) Strem# 46-1903. http://www.strem.com/catalog/v/46-1903/