Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 5g. Palladium, 5% on alumina powder, reduced, dry (Escat™ 1241). MFCD03613602
SDP
Catalog No. 5090112426
Encompass
CAS# 7440-05-3. 5g. Palladium, 5% on alumina powder, reduced, dry (Escat™ 1241). MFCD03613602. Molecular Weight: . Molecular Formula: . Color/form: gray pwdr. (d50=70 mum) Strem# 46-1951. http://www.strem.com/catalog/v/46-1951/
Catalog No. 50-901-12426 Supplier Strem, An Ascensus Company Supplier No. 4619515G
CAS# 7440-05-3. 5g. Palladium, 5% on alumina powder, reduced, dry (Escat™ 1241). MFCD03613602. Molecular Weight: . Molecular Formula: . Color/form: gray pwdr. (d50=70 mum) Strem# 46-1951. http://www.strem.com/catalog/v/46-1951/