Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 25g. Palladium, 5% on alumina, reduced, dry powder. MFCD03613602
SDP
Catalog No. 5090112424
Encompass
CAS# 7440-05-3. 25g. Palladium, 5% on alumina, reduced, dry powder. MFCD03613602. Molecular Weight: . Molecular Formula: Pd on alumina. Color/form: pwdr. Strem# 46-1950. http://www.strem.com/catalog/v/46-1950/
Catalog No. 50-901-12424 Supplier Strem, An Ascensus Company Supplier No. 46195025G
CAS# 7440-05-3. 25g. Palladium, 5% on alumina, reduced, dry powder. MFCD03613602. Molecular Weight: . Molecular Formula: Pd on alumina. Color/form: pwdr. Strem# 46-1950. http://www.strem.com/catalog/v/46-1950/