Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 50g. Palladium, 5% on barium sulfate, reduced. MFCD03613605
SDP
Catalog No. 5090112431
Encompass
CAS# 7440-05-3. 50g. Palladium, 5% on barium sulfate, reduced. MFCD03613605. Molecular Weight: . Molecular Formula: . Color/form: gray to black pwdr. Strem# 46-1989. http://www.strem.com/catalog/v/46-1989/
Catalog No. 50-901-12431 Supplier Strem, An Ascensus Company Supplier No. 46198950G
CAS# 7440-05-3. 50g. Palladium, 5% on barium sulfate, reduced. MFCD03613605. Molecular Weight: . Molecular Formula: . Color/form: gray to black pwdr. Strem# 46-1989. http://www.strem.com/catalog/v/46-1989/