Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 25g. Palladium, 5% on silica powder, reduced, dry (Escat™ 1351). MFCD06798745
SDP
Catalog No. 5090112456
Encompass
CAS# 7440-05-3. 25g. Palladium, 5% on silica powder, reduced, dry (Escat™ 1351). MFCD06798745. Molecular Weight: . Molecular Formula: . Color/form: gray pwdr. (d50=40 mum) Strem# 46-2090. http://www.strem.com/catalog/v/46-2090/
Catalog No. 50-901-12456 Supplier Strem, An Ascensus Company Supplier No. 46209025G
CAS# 7440-05-3. 25g. Palladium, 5% on silica powder, reduced, dry (Escat™ 1351). MFCD06798745. Molecular Weight: . Molecular Formula: . Color/form: gray pwdr. (d50=40 mum) Strem# 46-2090. http://www.strem.com/catalog/v/46-2090/