Learn More
Electron Microscopy Sciences Palladium/Gold 40, 0.20 mm, Purity 99.90% 50 FT
SDP
Catalog No. 501899991
Encompass_Preferred
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.90%
Catalog No. 50-189-9991 Supplier Electron Microscopy Sciences Supplier No. 73401
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.90%